Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Standard, APHA 10 Platinum-Cobalt, Hazen, Reagents
Click to view available options
Quantity:
500 mL
Specifications
Specifications
| Chemical Name or Material | Color Standard, APHA 10 Platinum-Cobalt, Hazen |
| Packaging | Glass Bottle |
| CAS | 7732-18-5, 7647-01-0 |
| Specific Gravity | 1.001 |
| Physical Form | Liquid |
| Color | Brown/Yellow |
| Boiling Point | 100°C |
| Solubility | Completely miscible |
| Quantity | 500 mL |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction