Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA 1000 Platinum-Cobalt, Hazen, Reagents

Catalog No. 01335654
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
01-335-654 500 mL
1 options

Catalog No. 01-335-654

Supplier: Reagents Holdings Llc CC115900500C

Only null left
Add to Cart
Add to Cart

Color Standard, APHA 1000 Platinum-Cobalt, Hazen, Reagent, use for Water and wastewater analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Color Standard, APHA 1000 Platinum-Cobalt, Hazen
Packaging Glass Bottle
CAS 7732-18-5, 7647-01-0, 16921-30-5
Specific Gravity 1.0243
Physical Form Liquid
Color Brown/Yellow
pH <1
Boiling Point 103°C
Solubility Completely miscible
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.