Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA 500 Platinum-Cobalt, Hazen, Reagents

Catalog No. 01335651
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-651 500 mL
01-335-653 120 mL
01-335-652 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-335-651 Supplier Reagents Holdings Llc Supplier No. CC116000500C
Only null left
Add to Cart
Add to Cart

Color Standard, APHA 500 Platinum-Cobalt, Hazen, Reagent, use for Water and wastewater analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Color Standard, APHA 500 Platinum-Cobalt, Hazen
Packaging Glass Bottle
CAS 7732-18-5, 7647-01-0, 16921-30-5
Specific Gravity 1.02
Physical Form Liquid
Color Brown/Yellow
pH <1
Boiling Point 103°C
Solubility Completely miscible
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.