Learn More
Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 1000, Ricca Chemical
Manufacturer:  Ricca Chemical Company 223116
Catalog No. 22-311-6
Add to cart
Specifications
Specifications
Color Standard
7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Amber glass
Colorless
APHA / Hazen (Platinum-Cobalt), Color = 1000
Liquid
Aqueous Solution
500mL
Documents
Promotions
Promotions
  • Ricca Chemical Company , Fisher Scientific , Fisherbrand
    Buy $250 of Ricca Chemical Company products and get $250 more to spend on Ricca items at no additional cost.
    06-30-2021