Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 25, Ricca Chemical
SDP

Catalog No. 223002516
Click to view available options
Quantity:
500 mL

Specifications

Chemical Name or Material Color Standard
Packaging Amber Glass Bottle
Name Note Color = 25
CAS 7732-18-5, 7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
CAS Min % 97.0, 0.5, 0.01, 0.0042
CAS Max % 100.0, 0.52, 0.01, 0.0044
Physical Form Liquid
Color Brown/Yellow to Yellow
pH <1
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type Color Reference Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.