Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 500, Ricca Chemical
$110.00 - $123.00
Specifications
Chemical Name or Material | Color Standard |
---|---|
Name Note | Color= 500 |
CAS Min % | 87.0, 10.32, 0.11, 0.09 |
CAS Max % | 91.0, 10.74, 0.12, 0.09 |
CAS | 16921-30-5, 7791-13-1, 7647-01-0 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
2230-16
|
Ricca Chemical Company
223016 |
500mL | Amber Glass |
Each for $419.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
2230-32
|
Ricca Chemical Company
223032 |
1L | Amber Glass |
Each for $713.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
22304
|
Ricca Chemical Company
22304 |
120mL | Amber Glass |
Each for $110.00
|
|
R2230000200
|
Ricca Chemical Company
2230200 |
200mL | Glass amber |
Each for $113.00
|
|
R2230000250
|
Ricca Chemical Company
22308 |
250mL | Glass amber |
Each for $123.00
|
|
Specifications
Color Standard | |
87.0, 10.32, 0.11, 0.09 | |
16921-30-5, 7791-13-1, 7647-01-0 |
Color= 500 | |
91.0, 10.74, 0.12, 0.09 | |
APHA/Hazen (Platinum-Cobalt) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title