Color Standard, Color No. 3, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 2230.00316

Quantity 500mL

 View more versions of this product

Catalog No. 223000316

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16921-30-5,7647-01-0,7732-18-5,7791-13-1
    Physical Form Liquid
    Chemical Name or Material APHA / Hazen (Platinum-Cobalt)

    Specifications

    Quantity 500mL
    Color Clear light yellow to yellow brown
    Physical Form Liquid
    CAS 7791-13-1,16921-30-5,7647-01-0,7732-18-5
    Chemical Name or Material APHA / Hazen (Platinum-Cobalt)
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit