Learn More
Color Standard CRA/CP 0.2 (RBU 5.0 SAP) for Corn Syrup Analysis, Reagents
Specifications
Specifications
| Chemical Name or Material | Color Standard CRA/CP 0.2 (RBU 5.0 SAP) for Corn Syrup Analysis |
| Packaging | Glass Bottle |
| CAS | 7732-18-5, 7647-01-0 |
| Specific Gravity | 1.021 |
| Physical Form | Liquid |
| Color | Yellow |
| Boiling Point | 100°C |
| Solubility | Completely miscible |
| Quantity | 100 mL |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.