Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Standard CRA/CP 0.5 (RBU 11.3 SAP) for Corn Syrup Analysis, Reagents
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Chemical Name or Material | Color Standard CRA/CP 0.5 (RBU 11.3 SAP) for Corn Syrup Analysis |
Packaging | Glass Bottle |
CAS | 7732-18-5, 7647-01-0 |
Specific Gravity | 1.021 |
Physical Form | Liquid |
Color | Yellow |
Boiling Point | 100°C |
Solubility | Completely miscible |
Quantity | 100 mL |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction