Learn More
US Biomax Inc COLORECTAL CANCER TISSUE ARRAY  DFS Item
Manufacturer:  US Biomax Inc BC000110
BC000110 : Colorectal cancer tissue array, including pathology grade, TNM and clinical stage, 85 cases/170 cores
Catalog No. NC1795537