Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Labeling Tape Available on GSA/VA Contract for Federal Government customers only.

Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.

$57.30 - $117.10

Specifications

Format Tape
Product Type Colored Label Tape
Quantity 6/Pk.
For Use With (Application) Waterproof, Oil and Acid resistant
Material Paper
View More Specs

Products 27
Catalog Number Color Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Color Length (English) Length (Metric) Price Quantity & Availability  
15-901-J
Accessories View Documents
Copper 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Add to cart
 
15-901-K
Accessories View Documents
Gray 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Add to cart
 
15-901-L Edge
Accessories View Documents
Lavender 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-M Edge
Accessories View Documents
Lime Green 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-N
Accessories View Documents
Rose 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Add to cart
 
15-901-Q
Accessories View Documents
Violet 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Add to cart
 
15-901-A Edge
Accessories View Documents
White 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-B Edge
Accessories View Documents
Yellow 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-C Edge
Accessories View Documents
Green 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-D Edge
Accessories View Documents
Pink 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-F Edge
Accessories View Documents
Orange 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-G Edge
Accessories View Documents
Blue 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-H Edge
Accessories View Documents
Aqua 14 yd. 13 m
Pack of 6 for $57.30
Case of 4 PK for $193.80
Edge
Add to cart
 
15-901-15J Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Copper 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15K Available on GSA/VA Contract for Federal Government customers only.
View Documents
Gray 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15L Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Lavender 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15N Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Rose 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15A Edge Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
White 60 yd. 55 m
Case of 6 for $117.10
Edge
Add to cart
 
15-901-15B Edge Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Yellow 60 yd. 55 m
Case of 6 for $117.10
Edge
Add to cart
 
15-901-15C Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Green 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15D Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Pink 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15E Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Red 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15F Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Orange 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15G Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Blue 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15H Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Aqua 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15M Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Lime Green 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
15-901-15Q Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Violet 60 yd. 55 m
Case of 6 for $117.10
Add to cart
 
Description

Description

Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.

  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
  • Tapes are supplied without dispenser
  • Core Size: 1 in. (14-yd. tape); 3 in. (60-yd. tape)
Specifications

Specifications

Tape
6/Pk.
Paper
-23°C to +121°C
0.5 in.
Colored Label Tape
Waterproof, Oil and Acid resistant
-10 to 250°F
13 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit