Learn More
Description
- Frosted writing surface
- Can be pierced for sample removal
Specifications
Specifications
| Includes | O-ring |
| Product Type | Screw Cap with O-ring |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Orange |
| Sterility | Non-sterile |
| For Use With (Equipment) | Skirted and Conical Bottom Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.