Learn More
ColorFlex™ Goggles
SCI_ED
Colorful, sparkling goggles are fun and chemical-splash resistant.
$30.60
Specifications
Certifications/Compliance ANSI Z87.1-1989 standards
Lens Tint Clear
Lens Type Anti-fog
Product Type Safety goggles
Frame Material PVC
View More Specs
Alert:
Straps contain some amount of latex.
Products 2
Catalog Number Mfr. No. Size Frame Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Frame Color Price Quantity & Availability  
S47617R
SCI_ED
View Documents Promotion Details
Oberon Company
7506
Regular Red
Case of 24 for $30.60
Save 50%
Reg : $61.21
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to cart
S47618B
SCI_ED
View Documents Promotion Details
Oberon Company
7406J
Junior Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Antifog-coated polycarbonate lens provides distortion-free vision
  • Fit over most glasses
  • Frame is made of flexible PVC for a comfortable fit
Compliance
Meet ANSI™ Z87.1-2003 (Z87+ High Impact).
Specifications
Specifications
ANSI Z87.1-1989 standards
Anti-fog
PVC
Polycarbonate
Clear
Safety goggles
Anti-fog