Learn More
ColorFlex™ Goggles Products for Science Education
Colorful, sparkling goggles are fun and chemical-splash resistant.
Manufacturer:  Oberon Company 7506
 View more versions of this product
Catalog No. S47617R
Add to cart
Alert:
Straps contain some amount of latex.
Description
Description
  • Antifog-coated polycarbonate lens provides distortion-free vision
  • Fit over most glasses
  • Frame is made of flexible PVC for a comfortable fit
Compliance
Meet ANSI™ Z87.1-2003 (Z87+ High Impact).
Specifications
Specifications
ANSI Z87.1-1989 standards
PVC
Polycarbonate
Clear
Safety goggles
Red
Anti-fog
Regular
Anti-fog
Education