Learn More
Electron Microscopy SciencesSupplier Diversity Partner Colormark™ Plus Glass Slide, White/Black Frosted
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 6340010
Encompass_Preferred
Specimen Slide identification is VITAL. Automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides. To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument. You need COLORMARK slides.
These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etched through the painted end on the front is clearly visible. Colormark Plus slides are adhesive coated therefore ensuring the adhesion of the section to the slide during staining procedures.
Label: White/Black Frosted
Catalog No. 50-250-56