Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ Colormark™ and Colormark™ Plus Microscope Slides Available on GSA/VA Contract for Federal Government customers only.
View etched patient information with higher visibility with the contrasting background color of Thermo Scientific™ Colormark™ Plus Adhesion Slides.
 View more versions of this product
Catalog No. 22-035-810
Add to cart
Specifications
Description
View etched patient information with higher visibility with the contrasting background color of Thermo Scientific™ Colormark™ Slides and Colormark Plus™ Adhesion Microscope Slides when used with the Thermo Scientific™ Shandon™ MicroWriter™ Slide Labeler and TBS Shur/Mark™ Slide Labeler. Colormark and Colormark Plus slides feature a 3/4 in. Superfrost or Colorfrost tab on the front with a contrasting dark tab on the back side of the slide so that the slides can be permanently etched with automated slide etching instruments. Colormark Plus Adhesion Microscope Slides hold tissue sections on permanently without the need for expensive coatings in IHC and Anatomical Pathology applications.
Designed for use with automated slide labeling machines. Offer excellent etched labelling visibility.
  • Self-contained packages ready for loading into your slide labeling system
  • Simply remove the cover from the slide package and drop the sleeve into the loading bin of the labeling system
  • Available with Plus formula to provide the utmost adhesion
Description & Specifications
Specifications
Colormark Plus
Plain
Colormark Plus