Learn More
Epredia™ Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Shop All Epredia Products
Click to view available options
Color:
Blue
Lilac/Black
Pink
White
Yellow
Coating:
Adhesion
Non-adhesion
Description
- Brightly coated with contrasting background color
- Self-contained packages ready for loading into your SHUR/Mark slide labeling system
- Simply remove the cover from the slide package and drop the sleeve into the loading bin of the SHUR/Mark labeling system
- Available with Plus formula to provide the utmost adhesion
Specifications
Specifications
| Color | Blue |
| Coating | Adhesion |
| Quantity | 144 Pk., 10 Pk./Cs. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov