Learn More
Thermo Scientific Chemicals ColorpHast™ pH Strips, Narrow, one square with single indicator on each strip, 11-13 pH range
Supplier:  Thermo Scientific Chemicals 035228LQ
 View more versions of this product
Catalog No. AA35228LQ
  • $70.65 / Each of 1
    Save 13%
    Reg : $81.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Order in Bulk
Request bulk or custom formats
Add to Cart
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
11 to 13
1 Unit (100 Strips)
Narrow pH Strip
Product Suggestions
Product Suggestions