Learn More
Thermo Scientific™ ColorStack Storage System
Achieve higher levels of organization when archiving slides with Thermo Scientific™ ColorStack Storage Systems.
$612.50 - $613.50
Specifications
Description 14-Drawer Cabinet
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
E4312CGRAY
View Documents
Thermo Scientific™
E43/12C/GREY
Gray
Each for $613.50
Add to cart
E4312CTAN
View Documents
Thermo Scientific™
E4312C/TAN
Tan
Each for $612.50
Add to cart
E4312CORANG
View Documents
Thermo Scientific™
E4312C/ORANGE
Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Achieve higher levels of organization when archiving slides with Thermo Scientific™ ColorStack Storage Systems. An effective visual method of sorting, these systems are a perfect compliment to the standard cabinets, and can be used with the LabStack cabinet series. The cabinet is cream epoxy-coated metal. The color-coded drawers have colored plastic fronts; available in 10 different shades.
Includes
  • 14 drawers of one color
  • Each of these storage drawers is 48 x 48 x 14cm and can hold 6,000 slides.
Compatablity
  • Fully compatible with all standard cabinets, bases, and options
  • Colorstack drawers can easily replace drawers in a standard 14-drawer cabinet
  • Special top cover is cream to match the ColorStack 14-Drawer Cabinet
Specifications
Specifications
14-Drawer Cabinet