Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Colour Solution for Aluminum, Reagents
Click to view available options
Quantity:
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Color Solution for Aluminum |
| CAS | 7732-18-5, 115-41-3 |
| Boiling Point | 100°C |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1 |
| Color | Yellow |
| Quantity | 1 L |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction