Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific? PEEK Fingertight Fittings.
Manufacturer:  Thermo Scientific™ U-467BLKX
 View more versions of this product
Catalog No. 03-170-447
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Ensure HPLC quality and robustness with Thermo Scientific? PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
Description & Specifications
Specifications
1/16in., 10-32 Coned, Delrin, Black
Column end plugs, Black
1 Each
LC Column