Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COM1/p8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | COM1/p8 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
COM1/p8 Polyclonal specifically detects COM1/p8 in Human samples. It is validated for Western Blot.Specifications
COM1/p8 | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Apoptosis | |
PBS with 0.02% Sodium Azide | |
26471 | |
Reacts with the N terminus of the human p8 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Candidate of metastasis 1, COM1P8, nuclear protein 1, nuclear protein, transcriptional regulator, 1, p8, Protein p8 | |
NUPR1 | |
IgG | |
Affinity Purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title