Learn More
Brady™ Combination Lockout Pouch Kit
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Combination Lockout Pouch Kit is a versatile lockout pouch contains useful electrical lockout devices and padlocks.
Manufacturer:  Brady™ 95552
Catalog No. 19-104-185
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical & mechanical risk lockouts
6 in.
2 in.
7 in.
Safety
Black
(1) lockout pouch; (1) multi-pole circuit breaker; (1) no-hole circuit breaker, 277v; (1) no-hole circuit breaker, 480/600v; (1) plug lockout; (1) gate valve lockout, 1-2.5 in.; (1) small ball valve lockout; (2) lockout tags
Canvas
2 in. x 7 in. x 6 in.