Learn More
Brady™ Combination Lockout Satchel with Brady Safety Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Combination Lockout Satchel is a convienent solution that includes everything needed to lockout the most common mechanical and electrical control points. Extra valve lockout devices are included.
Manufacturer:  Brady™ 99687
Catalog No. 19-104-205
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical & mechanical risk lockouts
14 in.
15 in.
4 in.
Safety
Black on red
(1) lockout satchel; (1) universal lockout w/ arm; (1) small ball valve lockout; (1) gate valve lockout, 1-2.5 in.; (1) gate valve lockout, 2.5-5 in.; (1) gate valve lockout, 5-6.5 in.; (1) single-pole circuit breaker; (1) no-hole circuit breaker, 277v; (1) no-hole circuit breaker, 480/600v; (1) multi-pole circuit breaker; (1) lockout cleat, 277v; (1) lockout cleat, 480/600v; (1) plug lockout, 220/550v; (1) wall switch; (2) kd safety padlocks, 1.5 in. shackle, red; (2) lockout tags; (1) all purpose cable lockout with 8 ft sheathed cable
Nylon
15 in. x 4 in. x 14 in.