Learn More
Brady™ Combination Lockout Toolbox with Brady Safety Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Combination Lockout Toolbox is a convienent kit that contains all of the basic tools needed to lock out gate and ball valves, circuit breakers, switches and plugs.
Manufacturer:  Brady™ 99684
Catalog No. 19-104-203
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical & mechanical risk lockouts
7.5 in.
8 in.
14 in.
Safety
Gray
(1) lockout box; (1) adjustable gate valve lockout; (1) small ball valve lockout; (1) large ball valve lockout; (1) single-pole circuit breaker lockout; (1) no-hole circuit breaker lockout, 277v; (1) no-hole circuit breaker lockout, 480/600v; (1) lockout cleat, 277v; (1) lockout cleat, 480/600v; (1) plug lockout, 110v; (1) wall switch lockout; (2) kd safety padlocks, 1.5 in. shackle, red; (2) lockout tags; (1) lockout pouch
Polyethylene
8 in. x 14 in. x 7.5 in.