Learn More
Brady™ Combined Lockout & Lock Box Station With Components, 6 Brady Safety Padlocks, 12 Tags
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Combined Lockout & Lock Box Station With Components is a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. Designed with the durability required for an industrial workplace.
Manufacturer:  Brady™ 99709
Catalog No. 19-104-037
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Text Legend: GROUP LPCKOUT
16 in.
6 in.
14 in.
Safety
Black on yellow
(12) lockout tag; (6) 1.5 in shackle red kd safety padlock; 1 to 2.5 in gate valve lockout; 1 to 6.5 in adjustable gate valve lockout; 110 v plug lockout; 120 v snap-on circuit breaker; 277 v clamp-on circuit breaker; 277 v lockout cleat; 3-in-1 plug lockout; 480/600 v lockout cleat; 490/600 v clamp-on circuit breaker; attachable lock box; lockout cabinet; multi-pole circuit breaker lockout; oversized circuit breaker; wall switch lockout
Powder-coated Powder-coated metal
6 in. x 14 in. x 16 in.