Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiSolvent Methanol-Free Solvent, MilliporeSigma™

CombiSolvent methanol-free solvent for volumetric Karl Fischer titration with one component reagents Aquastar™.

Manufacturer:  MilliporeSigma 1.88008.1038

Catalog No. M1880081038


Add to cart

Description

Description

Specifications

Specifications

0.80 g/cm3 (at 20°C)
1 L
Glass Bottle
12°C
5.8 to 6.3 (H2O, 20°C)
Safety and Handling

Safety and Handling

Recommended Storage : Store at 15 to 25°C

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit