Learn More
Combretastatin A4, Tocris Bioscience™
Antiangiogenic
Supplier:  R&D Systems 1495/5
 View more versions of this product
Catalog No. 14-955-RD
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Antitumor, antiangiogenic and antimetastatic agent, in vitro and in vivo. Inhibits tubulin polymerization.
Specifications
Specifications
2-Methoxy-5-[(1Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
>99%
C18H20O5
117048-59-6
5 mg
Observed MW: 316.35
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at -20°C