Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dentec Safety™ Comfort-Air™ 400NxMD Half-Mask Elastomeric Respirator with Filter Cover & Holder

White NxMD half-mask respirators feature a filter cover and holder and are NIOSH approved. OSHA silica dust compliant when used with a N95 or P100 filter. Provides protection against non-oil based particulate aerosols and COVID viruses.
$35.95
Specifications
Color | White |
---|---|
Material | Thermoplastic Elastomer |
Product Type | Half-Mask Respirator |
With Exhalation Valve | No |
Facepiece Material | Thermoplastic Elastomer |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-222-558
![]() |
Dentec Safety™
400NXMDSMNF |
Small/Medium |
Each for $35.95
|
|
|||||
19-222-559
![]() |
Dentec Safety™
400NXMDMLNF |
Medium/Large |
Each for $35.95
|
|
|||||
Description
- Facepiece low-profile design fits under industrial style and medical style face shields
- Extended field of vision
- No exhalation valve ensures the user's exhaled breath does not exit the mask
- Rolled sealing flange provides comfort and improves fitting capabilities
- Does not require user to adjust straps excessively tight
- Textured sealing area reduces risk of the mask slipping on the face
- White color allows for written personalization and also makes dirt or other markings more visible to simplify the cleaning and disinfecting process
- Designed to allow for the proper fitting of most safety & prescription eyewear and goggles
- Ultra light harness assembly provides additional comfort
- Cradle suspension fits comfortably under suspension systems for hard hats, grinding & welding helmets
- 8-position adjustment
Specifications
White | |
Half-Mask Respirator | |
Thermoplastic Elastomer | |
8-point |
Thermoplastic Elastomer | |
No | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title