Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ COMMD6 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results.
$344.00 - $976.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH7.0) containing 20% glycerol 0.15M NaCl, 1mM DTT |
Gene ID (Entrez) | 170622 |
Molecular Weight | TMW: 12kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25178105
|
Novus Biologicals™
NBP2517810.5MG |
0.5mg |
Each for $976.00
|
|
NBP25178101
|
Novus Biologicals™
NBP2517810.1MG |
0.1mg |
Each for $344.00
|
|
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-85 of Human COMMD6 The Recombinant Human COMMD6 Protein is derived from E. coli. The Recombinant Human COMMD6 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH7.0) containing 20% glycerol 0.15M NaCl, 1mM DTT | |
TMW: 12kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
COMMD6 | |
Yes |
SDS-Page | |
170622 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Acrg, Acrg Embryonic Lethality Minimal Region Ortholog, COMM Domain Containing 6, COMM Domain-Containing Protein 6 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title