Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548869
Description
COMMD9 Polyclonal antibody specifically detects COMMD9 in Human samples. It is validated for Western Blot.Specifications
COMMD9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9P000 | |
COMMD9 | |
KLH conjugated synthetic peptide between 147-176 amino acids from the C-terminal region of human COMMD9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
COMMD9 | |
1810029F08Rik; AA409293; COMM domain containing 9; COMM domain-containing protein 9; COMMD9; HSPC166; neuroprotective protein 9 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
29099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction