Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common beta Chain Human anti-Human, FITC, Clone: CSL311, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275914F

Catalog No. NBP275914F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Common beta Chain
CSL311
Flow Cytometry
Conjugated, Purified
CSF2RB
Human CD131, also called the cytokine receptor common subunit beta
Protein A purified
Phospho Specific
Primary
1439.0
Human
Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity)
Human
IgG4 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody binds to CD131.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit