Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common beta Chain Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270463AF405

Catalog No. N270463A405


Add to cart

Description

Description

Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Common beta Chain
OTI4H3
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CSF2RB
Full length recombinant protein of human Common beta Chain (NP_000386) produced in HEK293T cells.
Immunogen affinity purified
Phospho Specific
Primary
1439.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit