Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common beta Chain Antibody (OTI4H3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270463H
Description
Common beta Chain Monoclonal specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Common beta Chain | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CSF2RB | |
Full length recombinant protein of human Common beta Chain (NP_000386) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4H3 | |
Western Blot, Immunohistochemistry | |
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
Mouse | |
Affinity Purified | |
Phospho Specific | |
1439.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction