Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common beta Chain Mouse anti-Human, Clone: OTI4H3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270463
Description
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Common beta Chain | |
OTI4H3 | |
Unconjugated | |
Affinity Purified,Lyophilized | |
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
CSF2RB | |
Full length human recombinant protein of human CSF2RB (NP_000386) produced in HEK293T cell. | |
95.7 kDa | |
Primary | |
100 μg | |
Phospho Specific | |
Human |
Western Blot,Immunohistochemistry | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
1439 | |
Mouse | |
IgG1 | |
Monoclonal | |
Immunogen affinity purified | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title