Learn More
Common beta Chain Rat anti-Mouse, Clone: RM0112-42A7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122490
 View more versions of this product
Catalog No. NBP122490
Add to cart
Description
Description
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Common beta Chain
RM0112-42A7
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity)
Rat
IgG1
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-3 R Beta.
Mouse
Western Blot
LYOPH
Western Blot 1:500-1:1000
Purified
CSF2RB
Mouse recombinant protein of the extracellular domain of IL-3 R Beta
Protein G purified
RUO
Primary
1439
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.