Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Rat anti-Mouse, Alexa Fluor 750, Clone: 644346, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB7842S100UG
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry.Specifications
Common gamma Chain/IL-2 R gamma | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P34902 | |
IL2RG | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant mouse Common gamma Chain/IL-2 R gamma Trp23-Ala263 (predicted) Accession # P34902 | |
RUO | |
3561 | |
Mouse | |
IgG2b |
Flow Cytometry | |
644346 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
Rat | |
100 μg | |
Primary | |
Detects mouse Common gamma Chain/IL-2 R gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-2 R alpha , rmIL-2 R beta , recombinant human (rh) IL-2 R gamma , rhIL-15 R alpha , recombinant rat IL-2 R alpha , or recombinant canine IL-2 R alpha is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)