Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Compact Battery Powered Hydraulic Fork Lift Available on GSA/VA Contract for Federal Government customers only.

Allows easy movement in confined spaces and narrow aisles. Mopec™ Compact Battery Powered Hydraulic Fork Lift is a compact, battery powered lift designed to help alleviate physical strain caused by manual lifting and transporting.

Manufacturer:  Mopec JD305

Catalog No. 22-444-175


Add to cart

Description

Description

  • Can roll through any standard 78 in. (198.1cm) door
  • Lightweight, easy to operate, and highly maneuverable
  • Can lift all size bodies to most five tier storage racks
  • Has a built-in charger offering hundred of lifts on a single charge
  • The easy to read illuminated battery status voltmeter lets you know when it is time to recharge
  • Low-profile legs
  • This unique feature allows the technician to navigate this lift under virtually any stretcher or transport cart currently on the market
  • Eliminates the constant cranking motion of a manual lift

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

1 Year Warranty

Specifications

Specifications

Lift
Fits through a 198.1cm door
Battery Powered Hydraulic Fork
Fits through a 78 in. door
Battery powered
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit