Learn More
Thermo Scientific™ Compact Digital Microplate Shaker, 100-240V, 50/60Hz, US plug, PROMO
Harness the efficiency of our microplate shaker, capable of shaking up to four microplates in one run.
Manufacturer:  Thermo Scientific™ 8888002312N
Catalog No. 11676337FS
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • PID control ensures consistent shaking results and precise temperature control
  • Digital display for easy viewing
  • Compact design is ideal for placing in incubators (non-CO2), chambers, and refrigerators (up to 60°C)
  • Timer Range: Continuous or 1min to 99hrs 59min
  • Automatic Overload Shutdown: 4.4kg / 9.7lbs