Learn More
Thermo Scientific™ Compact Digital Mini Rotator, 100-240V, 50/60Hz, US plug, PROMO
Realize maximum flexiblity with the easy-to-use, digitally-controlled rotator that provides smooth, quiet horizontal mixing of different tube/vessel types.
Manufacturer:  Thermo Scientific™ 8888002512N
Catalog No. 11676339FS
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C). PID control ensures consistent and smooth rocking motion.
  • Digital Speed Control: 50 to 300rpm
  • Timer Range: Continuous or 1min to 99hrs 59min
Ordering Information:
A wide variety of accessories are available to customize unit for exact laboratory needs.