Learn More
Thermo Scientific™ Compact Digital Rocker, 100-240V, 50/60Hz, US plug, PROMO
Ensure gentle sample agitation with our compact, adjustable angle, digital display unit.
Manufacturer:  Thermo Scientific™ 8888001912N
Catalog No. 11676333FS
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C). PID control ensures consistent and smooth rocking motion.
  • Digital Speed Control: 5 to 100rpm
  • Easy to adjust tilt angle: 7° to 13°
  • Timer Range: Continuous or 1 min. to 99 hr., 59 min.
Includes:
Dimpled mat (tray, silicone pad, two securing bands).