Learn More
Macherey-Nagel Macherey-Nagel PF12 Plus.  DFS Item
Manufacturer:  Macherey-Nagel 919250
Macherey-Nagel PF12 Plus. Increased flexibility with More than 100 preprogrammed methods Nephelometric turbidity measurements Backlit graphic display with intuitive userguidance USB interface for data transfer, update and power supply
Catalog No. NC1671203