Learn More
Ohaus Compass CX Compact Scale
Designed for energy efficient weighing in labs, classrooms or the field.
Manufacturer: Ohaus 30428199
Description
- Up to 1,000 hours of continuous operation using standard alkaline batteries
- Large backlit display
- Stackable design.
Specifications
Backlit LCD | |
0.1 g | |
Stainless steel pan, AA batteries | |
1.5 sec | |
135 mm |
Compact Scale | |
41 mm | |
201 mm | |
0.51 kg | |
220 g |