Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3a Mouse anti-Human, Alexa Fluor™ 594, Clone: D17/1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223497AF594
Description
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
Complement C3a | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
C3 | |
Human C3a | |
Primary | |
Recognizes an epitope that is present on human C3, C3a and C3a (desArg). Does not cross-react with C4a or C5a. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
D17/1 | |
Western Blot, ELISA, Sandwich ELISA | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Mouse | |
0.1 mL | |
718 | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title