Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3d Mouse anti-Human, Clone: C3D/2891, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Complement C3d |
---|---|
Clone | C3D/2891 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Protein Array |
Applications | ELISA, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122956
|
Novus Biologicals
NBP30774120UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for ELISA, Protein Array.Specifications
Complement C3d | |
0.2 mg/ml | |
ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
10 mM PBS with 0.05% BSA | |
718 | |
IgG2b κ | |
Protein A or G purified |
C3D/2891 | |
ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
Recombinant full-length human Complement C3d protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title