Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C4b Mouse anti-Human, Alexa Fluor™ 532, Clone: 25.3F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP231554AF532

Catalog No. NP2314A32


Add to cart

Description

Description

Complement C4b Monoclonal antibody specifically detects Complement C4b in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Complement C4b
25.3F4
Western Blot, ELISA
basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Epitope specificity differs from that of NBP1-05084, as determined by inhibition ELISA.
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
C4B
C4 isolated from human plasma adsorbed onto aluminum hydroxide gel.
0.1 mL
Primary
721
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit