Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C5 Mouse anti-Human, Baboon, Alexa Fluor™ 750, Clone: C17/5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223498AF750

Catalog No. NP223498A70


Add to cart

Description

Description

Complement C5 Monoclonal antibody specifically detects Complement C5 in Human,Baboon samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Complement C5
C17/5
Western Blot, ELISA
anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone C17/5 reacts with human C5a/C5a(desArg) and cross-reacts with baboon C5a/C5a(desArg).
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
C5
Purified human C5a
0.1 mL
Primary
727
Baboon, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit