Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C5 Mouse anti-Human, Baboon, Clone: C17/5, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $493.00
Specifications
Antigen | Complement C5 |
---|---|
Clone | C17/5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223498
|
Novus Biologicals
NBP223498 |
0.25 mg |
Each for $493.00
|
|
NBP22349801
|
Novus Biologicals
NBP2234980.1ML |
0.1 mL |
Each for $378.00
|
|
NB019304
|
Novus Biologicals
NBP2234980.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Complement C5 Monoclonal specifically detects Complement C5 in Human, Baboon samples. It is validated for Western Blot, ELISA.Specifications
Complement C5 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human, Baboon | |
727 | |
Purified human C5a | |
Primary | |
Store at 4C. Do not freeze. |
C17/5 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Immunology, Innate Immunity, Stem Cell Markers | |
anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822 | |
C5 | |
IgG1 κ | |
Protein A or G purified | |
Clone C17/5 reacts with human C5a/C5a(desArg) and cross-reacts with baboon C5a/C5a(desArg). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title