Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C1r Mouse anti-Human, Canine, Biotin, Clone: OTI1F1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270318B

Catalog No. NBP270318B


Add to cart

Description

Description

Complement Component C1r Monoclonal antibody specifically detects Complement Component C1r in Human, Canine samples. It is validated for Western Blot.
Specifications

Specifications

Complement Component C1r
OTI1F1
Western Blot
Affinity Purified, Conjugated
C1R
Recombinant protein fragment corresponding to amino acids 262-562 of human Complement Component C1r (NP_001724) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Canine, Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41
Mouse
IgG2b
0.1 mL
Primary
715.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit