Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C1s Mouse anti-Human, Mouse, Rat, Monkey, Alexa Fluor™ 700, Clone: OTI2A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270327AF700

Catalog No. N270327A700


Add to cart

Description

Description

Complement Component C1s Monoclonal antibody specifically detects Complement Component C1s in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Complement Component C1s
OTI2A8
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
C1S
Full length recombinant protein of human Complement Component C1s (NP_001725) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human, Monkey, Mouse, Rat
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757
Mouse
IgG2a
0.1 mL
Primary
716.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit